4-Fluorobenzenesulfonamide ≥98.0%
Supplier: TCI America
F0953-25G
F0953-5G
TCF0953-25GEA
330.22
CAD
TCF0953-25G
TCF0953-5G
4-Fluorobenzenesulfonamide ≥98.0%
4-Fluorobenzenesulphonamide
CAS Number: 402-46-0
MDL Number: MFCD00025384
Molecular Formula: C6H6FNO2S
Molecular Weight: 175.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127
Formula:
C₆H₆FNO₂S Melting Pt: 127 °C |
MDL Number:
MFCD00025384 CAS Number: 402-46-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...