2-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0949-1G F0949-5G
TCF0949-1GEA 69.52 CAD
TCF0949-1G TCF0949-5G
2-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-m-toluic acid

CAS Number: 315-31-1
MDL Number: MFCD00042504
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 119


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-m-toluic Acid

Formula: C₈H₇FO₂
Melting Pt: 119 °C
MDL Number: MFCD00042504
CAS Number: 315-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=358c6ccdb95bfa921fd2b1511d882ff938408d1b&vid=358c6ccdb95bfa921fd2b1511d882ff938408d1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR