6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Paliperidone Related Compound B

F0944-1G F0944-5G
TCF0944-1GEA 50.56 CAD
TCF0944-1G TCF0944-5G
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride ≥98.0% (by HPLC, titration analysis)
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride

CAS Number: 84163-13-3
MDL Number: MFCD06658529
Molecular Formula: C12H13FN2O
Molecular Weight: 256.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C12H14ClFN2O
MW: 256.7000 g/mol
MDL Number: MFCD06658529
CAS Number: 84163-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4f2a2d156f51d13af6e99ebbf967a4e96a6eef3&vid=a4f2a2d156f51d13af6e99ebbf967a4e96a6eef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR