4-(3-Fluorobenzyloxy)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-((3-Fluorobenzyl)oxy)benzaldehyde

F0943-1G F0943-5G
TCF0943-1GEA 147.1 CAD
TCF0943-1G TCF0943-5G
4-(3-Fluorobenzyloxy)benzaldehyde ≥98.0% (by GC)
4-(3-Fluorobenzyloxy)benzaldehyde

CAS Number: 66742-57-2
MDL Number: MFCD02605317
Molecular Formula: C14H11FO2
Molecular Weight: 230.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 265
Melting point (°C): 48

Formula: C₁₄H₁₁FO₂
MW: 230.24 g/mol
Boiling Pt: 265 °C
Melting Pt: 48 °C
MDL Number: MFCD02605317
CAS Number: 66742-57-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05c2842c190eb90ddc7c6f87cce60f6f3860b87b&vid=05c2842c190eb90ddc7c6f87cce60f6f3860b87b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR