2-Fluoro-6-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0941-1G
F0941-5G
TCF0941-1GEA
44.24
CAD
TCF0941-1G
TCF0941-5G
2-Fluoro-6-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-6-methoxyphenylboronic acid
CAS Number: 78495-63-3
MDL Number: MFCD02179483
Molecular Formula: C7H8BFO3
Molecular Weight: 169.95
Form: Crystal
Color: White
Melting point (°C): 125
Other Product Names / Synonyms: 2-Fluoro-6-methoxybenzeneboronic Acid
Formula:
C₇H₈BFO₃ Melting Pt: 125 °C |
MDL Number:
MFCD02179483 CAS Number: 78495-63-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...