(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, Fmoc-Pip-OH, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, Fmoc-HoPro-OH, 1-Fmoc-L-pipecolic Acid, Fmoc-HomoPro-OH, (S)-1-Fmoc-2-piperidinecarboxylic Acid

F0938-1G F0938-5G
TCF0938-1GEA 64.78 CAD
TCF0938-1G TCF0938-5G
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid

CAS Number: 86069-86-5
MDL Number: MFCD00235898
Molecular Formula: C21H21NO4
Molecular Weight: 351.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 157
Specific rotation [a]20/D: -22 deg (C=1, MeOH)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (S)-1-Fmoc-2-piperidinecarboxylic Acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, 1-Fmoc-L-pipecolic Acid, Fmoc-Pip-OH

Formula: C₂₁H₂₁NO₄
MW: 351.40 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00235898
CAS Number: 86069-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52f7e8f0df88f1fa469cbf54c674209fa5a98ad1&vid=52f7e8f0df88f1fa469cbf54c674209fa5a98ad1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR