9-Fluorenylmethyl pentafluorophenyl carbonate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Carbonic Acid 9-Fluorenylmethyl Pentafluorophenyl Ester, Fmoc-OPfp, 9-Fluorenylmethyl Pentafluorophenyl Carbonate

F0936-1G F0936-5G
TCF0936-1GEA 69.52 CAD
TCF0936-1G TCF0936-5G
9-Fluorenylmethyl pentafluorophenyl carbonate ≥98.0% (by HPLC)
9-Fluorenylmethyl pentafluorophenyl carbonate

CAS Number: 88744-04-1
MDL Number: MFCD00013261
Molecular Formula: C21H11F5O3
Molecular Weight: 406.31
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 85
Lambda max.: 300 nm (EtOH)


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Fluorinated Building Blocks, Peptide Synthesis
Other Product Names / Synonyms: Carbonic Acid 9-Fluorenylmethyl Pentafluorophenyl Ester, Fmoc-OPfp

Formula: C₂₁H₁₁F₅O₃
MW: 406.31 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00013261
CAS Number: 88744-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea17a2124dbf0027deec568dc7ec12c7c23237d4&vid=ea17a2124dbf0027deec568dc7ec12c7c23237d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR