2-Fluoro-4-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid
F0935-1G
F0935-5G
TCF0935-1GEA
102.7
CAD
TCF0935-1G
TCF0935-5G
2-Fluoro-4-pyridineboronic acid (contains varying amounts of Anhydride)
2-Fluoro-4-pyridineboronic acid
CAS Number: 401815-98-3
MDL Number: MFCD04112345
Molecular Formula: C5H5BFNO2
Molecular Weight: 140.91
Form: Crystal
Color: White
Melting point (°C): 200
Other Product Names / Synonyms: 2-Fluoro-4-pyridylboronic Acid
Formula:
C₅H₅BFNO₂ MW: 140.91 Melting Pt: 200 °C |
MDL Number:
MFCD04112534 CAS Number: 401815-98-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...