2-Furaldehyde diethyl acetal ≥98.0% (by GC)

Supplier: TCI America

F0929-25G F0929-5G
TCF0929-25GEA 161.16 CAD
TCF0929-25G TCF0929-5G
2-Furaldehyde diethyl acetal ≥98.0% (by GC)
2-Furaldehyde diethyl acetal

CAS Number: 13529-27-6
MDL Number: MFCD00003245
Molecular Formula: C9H14O3
Molecular Weight: 170.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 64
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2-(Diethoxymethyl)furan, Furfural Diethyl Acetal

Formula: C₉H₁₄O₃
Boiling Pt: 81 °C
Density: 1.00 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00003245
CAS Number: 13529-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9714ca0e9b6c4f393268c90771ae6b394d1702fe&vid=9714ca0e9b6c4f393268c90771ae6b394d1702fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR