4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid ≥98.0%

Supplier: TCI America

Synonyms: Fmoc-4-Amb-OH, 4-(Fmoc-aminomethyl)benzoic Acid

F0925-1G F0925-5G
TCF0925-1GEA 81.05 CAD
TCF0925-1G TCF0925-5G
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid ≥98.0%
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid

CAS Number: 164470-64-8
MDL Number: MFCD01074693
Molecular Formula: C23H19NO4
Molecular Weight: 373.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 215
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-(Fmoc-aminomethyl)benzoic Acid

Formula: C₂₃H₁₉NO₄
MW: 373.41 g/mol
Melting Pt: 215 °C
MDL Number: MFCD01074693
CAS Number: 164470-64-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR