Febantel
Supplier: TCI America
Synonyms:
N-(((2-(2-MEthoxyacetamido)-4-(phenylsulphanyl)phenyl)amino)((methoxycarbonyl)imino)methyl)carbamate, N-(((2-(2-MEthoxyacetamido)-4-(phenylsulfanyl)phenyl)amino)((methoxycarbonyl)imino)methyl)carbamate
F0922-1G
F0922-5G
TCF0922-1GEA
59.57
CAD
TCF0922-1G
TCF0922-5G
Febantel
Febantel
CAS Number: 58306-30-2
MDL Number: MFCD01738527
Molecular Formula: C20H22N4O6S
Molecular Weight: 446.48
Melting point (°C): 130
Formula:
C₂₀H₂₂N₄O₆S MW: 446.48 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD01738527 CAS Number: 58306-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...