4-Fluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(4-Fluoro-3-methoxyphenyl)boronic acid, 4-Fluoro-3-methoxybenzeneboronic acid
F0919-1G
F0919-5G
TCF0919-1GEA
84.37
CAD
TCF0919-1G
TCF0919-5G
4-Fluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-3-methoxyphenylboronic acid
CAS Number: 854778-31-7
MDL Number: MFCD08056358
Molecular Formula: C7H8BFO3
Molecular Weight: 169.95
Form: Crystal
Color: White
Melting point (°C): 191
Other Product Names / Synonyms: 4-Fluoro-3-methoxybenzeneboronic Acid
Formula:
C₇H₈BFO₃ MW: 169.95 g/mol Melting Pt: 191 °C |
MDL Number:
MFCD08056358 CAS Number: 854778-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...