2-Fluoro-5-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2-Fluoro-5-methylbenzeneboronic acid, 2-Fluoro-5-methylboronic acid

F0915-1G F0915-5G
TCF0915-1GEA 63.2 CAD
TCF0915-1G TCF0915-5G
2-Fluoro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-5-methylphenylboronic acid

CAS Number: 166328-16-1
MDL Number: MFCD03428512
Molecular Formula: C7H8BFO2
Molecular Weight: 153.95
Form: Crystal
Color: White


Other Product Names / Synonyms: 2-Fluoro-5-methylbenzeneboronic Acid, 6-Fluoro-m-tolylboronic Acid

Formula: C₇H₈BFO₂
MW: 153.9466 g/mol
MDL Number: MFCD03428512
CAS Number: 166328-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ab63ccb3efa0028fa1145ffbfa830ec9c8a417a&vid=6ab63ccb3efa0028fa1145ffbfa830ec9c8a417a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR