2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Fmoc-Glycinol, N-Fmoc-ethanolamine, 2-(Fmoc-amino)-1-ethanol

F0912-25G F0912-5G
TCF0912-25GEA 396.58 CAD
TCF0912-25G TCF0912-5G
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol ≥98.0% (by HPLC, total nitrogen)
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol

CAS Number: 105496-31-9
MDL Number: MFCD00235927
Molecular Formula: C17H17NO3
Molecular Weight: 283.33
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 141
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(Fmoc-amino)-1-ethanol, N-Fmoc-ethanolamine

Formula: C₁₇H₁₇NO₃
MW: 283.33 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00235927
CAS Number: 105496-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcbce203042163eef27ae66edbba95ce378e2725&vid=bcbce203042163eef27ae66edbba95ce378e2725&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR