5-Fluoro-8-methoxyquinoline ≥98.0%
Supplier: TCI America
F0909-1G
F0909-5G
TCF0909-1GEA
143.78
CAD
TCF0909-1G
TCF0909-5G
5-Fluoro-8-methoxyquinoline ≥98.0%
5-Fluoro-8-methoxyquinoline
CAS Number: 439-88-3
MDL Number: MFCD18452297
Molecular Formula: C10H8FNO
Molecular Weight: 177.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 40
Storage Temperature: 0-10°C
Formula:
C₁₀H₈FNO Boiling Pt: 147 °C Melting Pt: 40 °C |
MDL Number:
MFCD18452297 CAS Number: 439-88-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...