4-(1H-1,2,4-Triazol-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
F0903-1G
F0903-5G
TCF0903-1GEA
252.8
CAD
TCF0903-1G
TCF0903-5G
4-(1H-1,2,4-Triazol-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)
4-(1H-1,2,4-Triazol-1-yl)benzaldehyde
CAS Number: 27996-86-7
MDL Number: MFCD02681969
Molecular Formula: C9H7N3O
Molecular Weight: 173.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 149
Other Product Names / Synonyms: 1-(4-Formylphenyl)-1,2,4-triazole
Formula:
C₉H₇N₃O Melting Pt: 149 °C |
MDL Number:
MFCD02681969 CAS Number: 27996-86-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...