4-Fluoro-D-phenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0901-1G F0901-5G
TCF0901-1GEA 102.7 CAD
TCF0901-1G TCF0901-5G
4-Fluoro-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
4-Fluoro-D-phenylalanine

CAS Number: 18125-46-7
MDL Number: MFCD00063063
Molecular Formula: C9H10FNO2
Molecular Weight: 183.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 247
Specific rotation [a]20/D: 24 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Phe(4-F)-OH

Formula: C₉H₁₀FNO₂
Melting Pt: 247 °C
MDL Number: MFCD00063063
CAS Number: 18125-46-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24b8fc642bbc037c0a53d158c28ae54928772691&vid=24b8fc642bbc037c0a53d158c28ae54928772691&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR