4-Fluoro-N-salicylideneaniline ≥98.0%

Supplier: TCI America

Synonyms: 2-(((4-Fluorophenyl)imino)methyl)phenol, 4-Fluoro-N-salicylalaniline

F0893-1G F0893-5G
TCF0893-1GEA 45.82 CAD
TCF0893-1G TCF0893-5G
4-Fluoro-N-salicylideneaniline ≥98.0%
4-Fluoro-N-salicylideneaniline

CAS Number: 3382-62-5
MDL Number: MFCD00020090
Molecular Formula: C13H10FNO
Molecular Weight: 215.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 84
Lambda max.: 349 nm (MeOH)


Product Categories: Synthetic Organic Chemistry, Ligands, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-N-salicylalaniline

Formula: C₁₃H₁₀FNO
MW: 215.23 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00020090
CAS Number: 3382-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6e9d16a356dbc9268a431ae8b9ff11e4618329f&vid=f6e9d16a356dbc9268a431ae8b9ff11e4618329f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR