4-Fluoro-N-(4-fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
F0892-1G
F0892-5G
TCF0892-1GEA
71.1
CAD
TCF0892-1G
TCF0892-5G
4-Fluoro-N-(4-fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)
4-Fluoro-N-(4-fluorobenzylidene)aniline
CAS Number: 39769-09-0
MDL Number: MFCD00017950
Molecular Formula: C13H9F2N
Molecular Weight: 217.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 65
Formula:
C₁₃H₉F₂N MW: 217.22 g/mol Melting Pt: 65 °C |
MDL Number:
MFCD00017950 CAS Number: 39769-09-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...