2-Fluoro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0886-1G F0886-5G
TCF0886-1GEA 206.98 CAD
TCF0886-1G TCF0886-5G
2-Fluoro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Fluoro-3-hydroxypyridine

CAS Number: 174669-74-0
MDL Number: MFCD04112569
Molecular Formula: C5H4FNO
Molecular Weight: 113.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 128


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-3-pyridinol

Formula: C₅H₄FNO
Melting Pt: 128 °C
MDL Number: MFCD04112569
CAS Number: 174669-74-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe10206334e758b7d3361c65f16a1326fb33c999&vid=fe10206334e758b7d3361c65f16a1326fb33c999&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR