3-Fluoro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0876-1G
F0876-5G
TCF0876-1GEA
54.51
CAD
TCF0876-1G
TCF0876-5G
3-Fluoro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-4-methoxyphenylboronic acid
CAS Number: 149507-26-6
MDL Number: MFCD00807404
Molecular Formula: C7H8BFO3
Molecular Weight: 169.95
Form: Crystal
Color: White
Melting point (°C): 198
Other Product Names / Synonyms: 3-Fluoro-4-methoxybenzeneboronic Acid
Formula:
C₇H₈BFO₃ Melting Pt: 198 °C |
MDL Number:
MFCD00807404 CAS Number: 149507-26-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...