(9H-Fluoren-9-yl)methyl hydrazinecarboxylate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0872-5G F0872-25G
TCF0872-5GEA 88.48 CAD
TCF0872-5G TCF0872-25G
(9H-Fluoren-9-yl)methyl hydrazinecarboxylate ≥98.0% (by HPLC, titration analysis)
(9H-Fluoren-9-yl)methyl hydrazinecarboxylate

CAS Number: 35661-51-9
MDL Number: MFCD00239419
Molecular Formula: C15H14N2O2
Molecular Weight: 254.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 171
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis
Other Product Names / Synonyms: Carbazic Acid 9-Fluorenylmethyl Ester, Fmoc-hydrazine, [(9H-Fluoren-9-ylmethoxy)carbonyl]hydrazine

Formula: C15H14N2O2
Melting Pt: 171 °C
MDL Number: MFCD00239419
CAS Number: 35661-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ae9f22a706122c540f26c0407073a331c2a0d66&vid=0ae9f22a706122c540f26c0407073a331c2a0d66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR