Formononetin ≥98.0%

Supplier: TCI America

Synonyms: 7-Hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 7-Hydroxy-3-(4-methoxyphenyl)chromone, 7-Hydroxy-4'-methoxyisoflavone

F0868-1G F0868-5G
TCF0868-1GEA 198.29 CAD
TCF0868-1G TCF0868-5G
Formononetin ≥98.0%
Formononetin

CAS Number: 485-72-3
MDL Number: MFCD00016948
Molecular Formula: C16H12O4
Molecular Weight: 268.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 258
Lambda max.: 300 nm (EtOH)


Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 7-Hydroxy-3-(4-methoxyphenyl)chromone, 7-Hydroxy-4'-methoxyisoflavone

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
Melting Pt: 258 °C
MDL Number: MFCD00016948
CAS Number: 485-72-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df66d11643275b74446b293a58290e7bcc12ac1a&vid=df66d11643275b74446b293a58290e7bcc12ac1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR