1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Fmoc-cycloleucine, Fmoc-Cle-OH, 1-(Fmoc-amino)cyclopentanecarboxylic Acid

F0866-1G F0866-5G
TCF0866-1GEA 145.36 CAD
TCF0866-1G TCF0866-5G
1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid ≥95.0% (by HPLC, titration analysis)
1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid

CAS Number: 117322-30-2
MDL Number: MFCD01074696
Molecular Formula: C21H21NO4
Molecular Weight: 351.40
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 187
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-(Fmoc-amino)cyclopentanecarboxylic Acid, Fmoc-cycloleucine

Formula: C₂₁H₂₁NO₄
MW: 351.40 g/mol
Melting Pt: 187 °C
MDL Number: MFCD01074696
CAS Number: 117322-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cd3d7d27687c23dbba313449e4a63f0104f6f75&vid=8cd3d7d27687c23dbba313449e4a63f0104f6f75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR