4-Fluoro-D-2-phenylglycine ≥98.0%

Supplier: TCI America

Synonyms: H-D-Phg(4-F)-OH, (R)-2-Amino-2-(4-fluorophenyl)acetic Acid

F0863-1G F0863-5G
TCF0863-1GEA 158 CAD
TCF0863-1G TCF0863-5G
4-Fluoro-D-2-phenylglycine ≥98.0%
4-Fluoro-D-2-phenylglycine

CAS Number: 93939-74-3
MDL Number: MFCD00042727
Molecular Formula: C8H8FNO2
Molecular Weight: 169.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -135 deg (C=1, 1mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-2-Amino-2-(4-fluorophenyl)acetic Acid, H-D-Phg(4-F)-OH

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
MDL Number: MFCD00042727
CAS Number: 93939-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2b0e797ee84d25c44e32ce78ed82bc6bf3fefe5&vid=d2b0e797ee84d25c44e32ce78ed82bc6bf3fefe5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR