4-Fluoro-L-2-phenylglycine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0862-1G F0862-5G
TCF0862-1GEA 48.98 CAD
TCF0862-1G TCF0862-5G
4-Fluoro-L-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
4-Fluoro-L-2-phenylglycine

CAS Number: 19883-57-9
MDL Number: MFCD00213799
Molecular Formula: C8H8FNO2
Molecular Weight: 169.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 138 deg (C=1, 1mol/L Hcl)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-2-(4-fluorophenyl)acetic Acid, H-Phg(4-F)-OH

Formula: C₈H₈FNO₂
MDL Number: MFCD00213799
CAS Number: 19883-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76248df70accb513df4094071eb3bf37837b17fe&vid=76248df70accb513df4094071eb3bf37837b17fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR