5-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0859-1G
F0859-5G
TCF0859-1GEA
170.32
CAD
TCF0859-1G
TCF0859-5G
5-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
5-Fluoro-3-pyridineboronic acid
CAS Number: 872041-86-6
MDL Number: MFCD07368243
Molecular Formula: C5H5BFNO2
Molecular Weight: 140.91
Form: Crystal
Color: White
Melting point (°C): 236
Other Product Names / Synonyms: 5-Fluoro-3-pyridylboronic Acid
Formula:
C₅H₅BFNO₂ Melting Pt: 236 °C |
MDL Number:
MFCD07368243 CAS Number: 872041-86-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...