N-Formylsaccharin ≥98.0%

Supplier: TCI America

Synonyms: 3-Oxobenzo[d]isothiazole-2(3H)-carbaldehyde 1,1-dioxide

F0854-25G F0854-5G
TCF0854-25GEA 447.14 CAD
TCF0854-25G TCF0854-5G
N-Formylsaccharin ≥98.0%
N-Formylsaccharin

CAS Number: 50978-45-5
Molecular Formula: C8H5NO4S
Molecular Weight: 211.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₈H₅NO₄S
MDL Number: MFCD23380208
CAS Number: 50978-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e99ed09c9b7bf1415c123d88abd5376441c4ed9e&vid=e99ed09c9b7bf1415c123d88abd5376441c4ed9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR