Fenclorim ≥98.0% (by GC)

Supplier: TCI America

F0851-1G F0851-5G
TCF0851-1GEA 128.93 CAD
TCF0851-1G TCF0851-5G
Fenclorim ≥98.0% (by GC)
Fenclorim

CAS Number: 3740-92-9
MDL Number: MFCD00754193
Molecular Formula: C10H6Cl2N2
Molecular Weight: 225.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 97


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: 4,6-Dichloro-2-phenylpyrimidine

Formula: C₁₀H₆Cl₂N₂
Melting Pt: 97 °C
MDL Number: MFCD00754193
CAS Number: 3740-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28685221f9c2db1faf8cc018cdfc959c0fb25323&vid=28685221f9c2db1faf8cc018cdfc959c0fb25323&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR