3-Fluoro-4-morpholinoaniline ≥98.0%

Supplier: TCI America

Synonyms: 4-(2-Fluoro-4-aminophenyl)morpholine

F0849-25G F0849-5G
TCF0849-25GEA 426.6 CAD
TCF0849-25G TCF0849-5G
3-Fluoro-4-morpholinoaniline ≥98.0%
3-Fluoro-4-morpholinoaniline

CAS Number: 93246-53-8
MDL Number: MFCD02571270
Molecular Formula: C10H13FN2O
Molecular Weight: 196.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(2-Fluoro-4-aminophenyl)morpholine

Formula: C10H13FN2O
Melting Pt: 123 °C
MDL Number: MFCD02571270
CAS Number: 93246-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73f2f53b3d0d7c0d4a9a1347c1e28c1e7e8aa8d6&vid=73f2f53b3d0d7c0d4a9a1347c1e28c1e7e8aa8d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR