5-Fluoro-o-toluic acid ≥98.0%

Supplier: TCI America

F0844-25G F0844-5G
TCF0844-25GEA 279.66 CAD
TCF0844-25G TCF0844-5G
5-Fluoro-o-toluic acid ≥98.0%
5-Fluoro-o-toluic acid

CAS Number: 33184-16-6
MDL Number: MFCD00042294
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-o-toluic Acid

Formula: C₈H₇FO₂
Melting Pt: 133 °C
MDL Number: MFCD00042294
CAS Number: 33184-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=593a84b4a2855292014de40d504ffea753b3108f&vid=593a84b4a2855292014de40d504ffea753b3108f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR