3'-Fluoro-4'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)

Supplier: TCI America

F0843-1G F0843-5G
TCF0843-1GEA 94.8 CAD
TCF0843-1G TCF0843-5G
3'-Fluoro-4'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)
3'-Fluoro-4'-hydroxyacetophenone

CAS Number: 403-14-5
MDL Number: MFCD00215840
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 127

Formula: C₈H₇FO₂
Melting Pt: 127 °C
MDL Number: MFCD00215840
CAS Number: 403-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=900fd3f5d9f059e9a9b2c2f4e5e79da558c9cdb3&vid=900fd3f5d9f059e9a9b2c2f4e5e79da558c9cdb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR