5-Fluoropicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0838-1G F0838-5G
TCF0838-1GEA 74.26 CAD
TCF0838-1G TCF0838-5G
5-Fluoropicolinic acid ≥98.0% (by GC, titration analysis)
5-Fluoropicolinic acid

CAS Number: 107504-08-5
MDL Number: MFCD04114196
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 165


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoropicolinic Acid

Formula: C₆H₄FNO₂
Melting Pt: 165 °C
MDL Number: MFCD04114196
CAS Number: 107504-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a38b1eef60ac6cc7b0e31c380f6a5b183f90512c&vid=a38b1eef60ac6cc7b0e31c380f6a5b183f90512c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR