6-Fluoropicolinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0837-1G F0837-5G
TCF0837-1GEA 87.53 CAD
TCF0837-1G TCF0837-5G
6-Fluoropicolinic acid ≥98.0% (by titrimetric analysis)
6-Fluoropicolinic acid

CAS Number: 402-69-7
MDL Number: MFCD02181193
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 138


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoropicolinic Acid

Formula: C₆H₄FNO₂
Melting Pt: 138 °C
MDL Number: MFCD02181193
CAS Number: 402-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3695ad0e2e624ea6c14c6e410f4aa5805a2c1e9&vid=a3695ad0e2e624ea6c14c6e410f4aa5805a2c1e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR