5-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0834-1G
F0834-5G
TCF0834-1GEA
69.52
CAD
TCF0834-1G
TCF0834-5G
5-Formyl-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
5-Formyl-2-methoxyphenylboronic acid
CAS Number: 127972-02-5
MDL Number: MFCD01319044
Molecular Formula: C8H9BO4
Molecular Weight: 179.97
Form: Crystal
Color: Slightly Pale Yellow
Other Product Names / Synonyms: 5-Formyl-2-methoxybenzeneboronic Acid
Formula:
C₈H₉BO₄ |
MDL Number:
MFCD01319044 CAS Number: 127972-02-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...