6-Fluoro-4-chromanone ≥98.0% (by GC)

Supplier: TCI America

F0831-1G F0831-5G
TCF0831-1GEA 66.04 CAD
TCF0831-1G TCF0831-5G
6-Fluoro-4-chromanone ≥98.0% (by GC)
6-Fluoro-4-chromanone

CAS Number: 66892-34-0
MDL Number: MFCD00038654
Molecular Formula: C9H7FO2
Molecular Weight: 166.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 116

Formula: C₉H₇FO₂
Melting Pt: 116 °C
MDL Number: MFCD00038654
CAS Number: 66892-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b93433c58ec5bfd6082301e0bf37f2e7ebb27c11&vid=b93433c58ec5bfd6082301e0bf37f2e7ebb27c11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR