4-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0830-1G
F0830-5G
TCF0830-1GEA
26.86
CAD
TCF0830-1G
TCF0830-5G
4-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-3-(trifluoromethyl)phenylboronic acid
CAS Number: 182344-23-6
MDL Number: MFCD07363787
Molecular Formula: C7H5BF4O2
Molecular Weight: 207.92
Form: Crystal
Color: White
Melting point (°C): 174
Other Product Names / Synonyms: 4-Fluoro-3-(trifluoromethyl)benzeneboronic Acid
Formula:
C₇H₅BF₄O₂ Melting Pt: 174 °C |
MDL Number:
MFCD07363787 CAS Number: 182344-23-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...