4-Fluoro-o-anisaldehyde ≥97.0%

Supplier: TCI America

Synonyms: 4-Fluoro-2-methoxybenzaldehyde, 4-Fluoro-2-anisaldehyde

F0827-1G F0827-5G
TCF0827-1GEA 64.78 CAD
TCF0827-1G TCF0827-5G
4-Fluoro-o-anisaldehyde ≥97.0%
4-Fluoro-o-anisaldehyde

CAS Number: 450-83-9
MDL Number: MFCD00143318
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 61


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-2-methoxybenzaldehyde

Formula: C₈H₇FO₂
Melting Pt: 61 °C
MDL Number: MFCD00143318
CAS Number: 450-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41238893ac07a91d13445c0c78d8d1553e00f67b&vid=41238893ac07a91d13445c0c78d8d1553e00f67b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR