6-Fluoro-o-toluidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0817-1G F0817-5G
TCF0817-1GEA 41.08 CAD
TCF0817-1G TCF0817-5G
6-Fluoro-o-toluidine ≥98.0% (by titrimetric analysis)
6-Fluoro-o-toluidine

CAS Number: 443-89-0
MDL Number: MFCD06658252
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 77
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-3-fluorotoluene, 6-Fluoro-o-toluidine

Formula: C₇H₈FN
Boiling Pt: 89 °C
Density: 1.12 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD06658252
CAS Number: 443-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3815620428ac3cc79f51ac4cb0693c2c4d9f1d5&vid=a3815620428ac3cc79f51ac4cb0693c2c4d9f1d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR