2-Fluoro-4-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
2-Fluoro-4-(trifluoromethyl)benzeneboronic acid
F0816-1G
F0816-5G
TCF0816-1GEA
135.88
CAD
TCF0816-1G
TCF0816-5G
2-Fluoro-4-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-4-(trifluoromethyl)phenylboronic acid
CAS Number: 503309-11-3
MDL Number: MFCD03094995
Molecular Formula: C7H5BF4O2
Molecular Weight: 207.92
Form: Crystal
Color: White
Melting point (°C): 116
Other Product Names / Synonyms: 2-Fluoro-4-(trifluoromethyl)benzeneboronic Acid
Formula:
C₇H₅BF₄O₂ MW: 207.92 g/mol Melting Pt: 116 °C |
MDL Number:
MFCD03094995 CAS Number: 503309-11-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...