1-Fluoro-3-iodo-5-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

F0815-1G F0815-5G
TCF0815-1GEA 127.98 CAD
TCF0815-1G TCF0815-5G
1-Fluoro-3-iodo-5-nitrobenzene ≥98.0% (by GC)
1-Fluoro-3-iodo-5-nitrobenzene

CAS Number: 3819-88-3
MDL Number: MFCD00007219
Molecular Formula: C6H3FINO2
Molecular Weight: 267.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79

Formula: C₆H₃FINO₂
Melting Pt: 79 °C
MDL Number: MFCD00007219
CAS Number: 3819-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d79de6c31d614d591004560afd22b2008b7deb79&vid=d79de6c31d614d591004560afd22b2008b7deb79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR