2-Fluoro-p-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0813-1G F0813-5G
TCF0813-1GEA 69.52 CAD
TCF0813-1G TCF0813-5G
2-Fluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-p-toluic acid

CAS Number: 7697-23-6
MDL Number: MFCD03092906
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 186


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-p-toluic Acid

Formula: C₈H₇FO₂
Melting Pt: 186 °C
MDL Number: MFCD03092906
CAS Number: 7697-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=771973e500ae2d110f14a4d503e3241dfc5e4cba&vid=771973e500ae2d110f14a4d503e3241dfc5e4cba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR