4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97.0%
Supplier: TCI America
Synonyms:
1-(4-Fluoro-3-(trifluoromethyl)phenyl)ethanone
F0808-1G
F0808-5G
TCF0808-1GEA
94.17
CAD
TCF0808-1G
TCF0808-5G
4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97.0%
4'-Fluoro-3'-(trifluoromethyl)acetophenone
CAS Number: 208173-24-4
MDL Number: MFCD00061252
Molecular Formula: C9H6F4O
Molecular Weight: 206.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.36
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Acetyl-2-fluorobenzotrifluoride
Formula:
C₉H₆F₄O MW: 206.14 g/mol Density: 1.36 (20 °C) |
MDL Number:
MFCD00061252 CAS Number: 208173-24-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...