9-Fluorenone-4-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 9-Oxofluoren-4-carboxylic acid, 9-Oxo-9H-fluorene-4-carboxylic acid, 9-Oxofluorene-4-carboxylic acid

F0804-1G F0804-5G
TCF0804-1GEA 42.66 CAD
TCF0804-1G TCF0804-5G
9-Fluorenone-4-carboxylic acid ≥98.0%
9-Fluorenone-4-carboxylic acid

CAS Number: 6223-83-2
MDL Number: MFCD00001145
Molecular Formula: C14H8O3
Molecular Weight: 224.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228

Formula: C₁₄H₈O₃
Melting Pt: 228 °C
MDL Number: MFCD00001145
CAS Number: 6223-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbb564542384b391e36f1dea8d5eac6f0b044137&vid=fbb564542384b391e36f1dea8d5eac6f0b044137&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR