2-Fluoro-4-nitroaniline ≥98.0% (by GC)

Supplier: TCI America

F0798-1G F0798-5G
TCF0798-1GEA 37.92 CAD
TCF0798-1G TCF0798-5G
2-Fluoro-4-nitroaniline ≥98.0% (by GC)
2-Fluoro-4-nitroaniline

CAS Number: 369-35-7
MDL Number: MFCD00034560
Molecular Formula: C6H5FN2O2
Molecular Weight: 156.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 135

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00034560
CAS Number: 369-35-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8eb97d3f590d81d71d13c6f6991273348140388&vid=b0232daacad9d1fff0c190ddb10824e985fde8e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR