2-Fluoro-4-nitrobenzoic acid ≥98.0%

Supplier: TCI America

F0797-25G F0797-5G
TCF0797-25GEA 290.88 CAD
TCF0797-25G TCF0797-5G
2-Fluoro-4-nitrobenzoic acid ≥98.0%
2-Fluoro-4-nitrobenzoic acid

CAS Number: 403-24-7
MDL Number: MFCD00275565
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00275565
CAS Number: 403-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b90698bc7f15d036d112ce3b45d29a19bbad1f74&vid=df946a267b3f9ff19bb50c253556f10b97005781&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR