2-Fluoro-6-iodobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0796-1G F0796-5G
TCF0796-1GEA 42.66 CAD
TCF0796-1G TCF0796-5G
2-Fluoro-6-iodobenzoic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-6-iodobenzoic acid

CAS Number: 111771-08-5
MDL Number: MFCD00042289
Molecular Formula: C7H4FIO2
Molecular Weight: 266.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 126

Formula: C₇H₄FIO₂
Melting Pt: 126 °C
MDL Number: MFCD00042289
CAS Number: 111771-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=408ae7b8d94f04be0a16b6182bfe2f480d0d6259&vid=22ecc7b445faf49afaadd5b7ff094f408747ea70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR