4-Fluoro-2-methylbenzaldehyde ≥95.0% (by GC)
Supplier: TCI America
F0793-1G
F0793-5G
TCF0793-1GEA
33.18
CAD
TCF0793-1G
TCF0793-5G
4-Fluoro-2-methylbenzaldehyde ≥95.0% (by GC)
4-Fluoro-2-methylbenzaldehyde
CAS Number: 63082-45-1
MDL Number: MFCD02261764
Molecular Formula: C8H7FO
Molecular Weight: 138.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 55
Flash Point (°C): 82
Specific Gravity (20/20): 1.16
Formula:
C₈H₇FO Boiling Pt: 55 °C Density: 1.16 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD02261764 CAS Number: 63082-45-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...