5-Fluoro-2-nitroaniline ≥98.0%

Supplier: TCI America

F0789-25G F0789-5G
TCF0789-25GEA 412.38 CAD
TCF0789-25G TCF0789-5G
5-Fluoro-2-nitroaniline ≥98.0%
5-Fluoro-2-nitroaniline

CAS Number: 2369-11-1
MDL Number: MFCD00034065
Molecular Formula: C6H5FN2O2
Molecular Weight: 156.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98

Formula: C₆H₅FN₂O₂
Melting Pt: 98 °C
MDL Number: MFCD00034065
CAS Number: 2369-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40c0758a05a844a75a9b6109f42a7d7ea004c117&vid=40c0758a05a844a75a9b6109f42a7d7ea004c117&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR