2-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0788-1G
F0788-5G
TCF0788-1GEA
145.36
CAD
TCF0788-1G
TCF0788-5G
2-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-5-(trifluoromethyl)phenylboronic acid
CAS Number: 352535-96-7
MDL Number: MFCD03701538
Molecular Formula: C7H5BF4O2
Molecular Weight: 207.92
Form: Crystal
Color: White
Melting point (°C): 109
Other Product Names / Synonyms: 2-Fluoro-5-(trifluoromethyl)benzeneboronic Acid
Formula:
C₇H₅BF₄O₂ Melting Pt: 109 °C |
MDL Number:
MFCD03701538 CAS Number: 352535-96-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...